Learn More
Research Products International Corp Inoculating Loop, 10ul, YLW, S, Ind. Wrap., 250/cs
SDP
Catalog No. 502229826
Encompass
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polystyrene for easy disposal after each use. Supplied in sealed packs with certification. Individually wrapped, 250 per case.
Catalog No. 50-222-9826 Supplier Research Products International Corp Supplier No. 1612121S
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polystyrene for easy disposal after each use. Supplied in sealed packs with certification. Individually wrapped, 250 per case.