Learn More
Chemglass Life Sciences Inoculation Pick, Large, Semi Point, 0.3175cm OD x 15.24cm Long
SDP
Catalog No. 501311332
Encompass_Preferred
Inoculation Pick, Large, Semi Point, 0.3175cm OD x 15.24cm Long. • Go from autoclave to microtube• Available in three lengths and with semi or needle points• Non-sterile, meet FDA requirements for food• 100% USA grown White Birch• Environmentally friendly – naturally biodegradableMade in a USA, ISO and FDA registered facility, wooden inoculation picks provide a faster way to inoculate. It’s simple, after autoclaving, just “pick” the colony, drop the wooden pick into your media and close the lid. No need to remove the pick. Available in three convenient sizes to accommodate various transfer, inoculation and streaking needs.
Catalog No. 50-131-1332 Supplier Chemglass Life Sciences Supplier No. CLS3670108
Inoculation Pick, Large, Semi Point, 0.3175cm OD x 15.24cm Long. • Go from autoclave to microtube• Available in three lengths and with semi or needle points• Non-sterile, meet FDA requirements for food• 100% USA grown White Birch• Environmentally friendly – naturally biodegradableMade in a USA, ISO and FDA registered facility, wooden inoculation picks provide a faster way to inoculate. It’s simple, after autoclaving, just “pick” the colony, drop the wooden pick into your media and close the lid. No need to remove the pick. Available in three convenient sizes to accommodate various transfer, inoculation and streaking needs.