Learn More
Cole-Parmer Argos Technologies Inoculating Needles, 3", 25 Gauge; 12/Pk
Catalog No. 501960663 Shop All Cole-Parmer Products
Encompass_Preferred
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.
Catalog No. 50-196-0663 Supplier Cole-Parmer Supplier No. 0439649
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.