Learn More
Cole-Parmer Argos Technologies Calibrated Inoculating Loops, 10 uL, 4 mm ID; 5/Pk
Catalog No. 501960599 Shop All Cole-Parmer Products
Encompass_Preferred
Argos brand Inoculating Loops are made from high quality twisted nichrome wire. The loops have been designed to slide along agar surfaces without cutting or tearing. They cool rapidly and are less expensive than platinum. The loops are supplied without handles. They are available calibrated or non-calibrated. Brass loop handles are sold separately as models, 04396-40 and -41.
Catalog No. 50-196-0599 Supplier Cole-Parmer Supplier No. 0439635
Argos brand Inoculating Loops are made from high quality twisted nichrome wire. The loops have been designed to slide along agar surfaces without cutting or tearing. They cool rapidly and are less expensive than platinum. The loops are supplied without handles. They are available calibrated or non-calibrated. Brass loop handles are sold separately as models, 04396-40 and -41.