Learn More
Cole-Parmer Argos Technologies Medium Inoculating Loops, 3.41 mm ID; 25/Pk
Catalog No. 501960653 Shop All Cole-Parmer Products
Encompass_Preferred
These inoculating loops are made from high quality twisted nichrome wire. The loops have been designed to slide along agar surfaces without cutting or tearing. They cool rapidly and are less expensive than platinum. The loops are supplied without handles and available calibrated or non-calibrated. Brass loop handles are sold separately as models 04396-40 and -41.
Catalog No. 50-196-0653 Supplier Cole-Parmer Supplier No. 0439638
These inoculating loops are made from high quality twisted nichrome wire. The loops have been designed to slide along agar surfaces without cutting or tearing. They cool rapidly and are less expensive than platinum. The loops are supplied without handles and available calibrated or non-calibrated. Brass loop handles are sold separately as models 04396-40 and -41.