Learn More
Electron Microscopy Sciences Inoculating Needles, Rigid
SDP
Catalog No. 501929180
Encompass_Preferred
Sterile, disposable and verifiably accurate plastic loops and needles are designed for use in quantitative procedures such as sampling, urine counts and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and single use, there is no risk of cross-contamination and no need for flaming. They are flexible enough for streaking, yet rigid enough for subsurface inoculation.
The unique hexaginal shape of the shaft makes it easier to grip, position and guide through a smooth streaking process. The loop surfaces are extremely smooth, with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface. These loops provide an efficient, cost-effective and time saving alternative to traditional metallic loops.
Material: Rigid
Length: 200 mm
Catalog No. 50-192-9180 Supplier Electron Microscopy Sciences Supplier No. 6244403
Sterile, disposable and verifiably accurate plastic loops and needles are designed for use in quantitative procedures such as sampling, urine counts and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and single use, there is no risk of cross-contamination and no need for flaming. They are flexible enough for streaking, yet rigid enough for subsurface inoculation.
The unique hexaginal shape of the shaft makes it easier to grip, position and guide through a smooth streaking process. The loop surfaces are extremely smooth, with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface. These loops provide an efficient, cost-effective and time saving alternative to traditional metallic loops.
Material: Rigid
Length: 200 mm