Learn More
CELLTREAT Scientific Products 10l Inoculating Loop, Individual Pack, Sterile
SDP
Catalog No. 50201985
Encompass_Preferred
CELLTREAT Scientific Products 10¿L Inoculating Loop. Flexible for easy access to small volume tubes and dishes. Color coded blue for easy identification. Gentle on delicate agar or other surfaces. Available in peel-to-open paper/plastic wrappers. 221mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 600/case
Catalog No. 50-201-985 Supplier CELLTREAT Scientific Products Supplier No. 229612
CELLTREAT Scientific Products 10¿L Inoculating Loop. Flexible for easy access to small volume tubes and dishes. Color coded blue for easy identification. Gentle on delicate agar or other surfaces. Available in peel-to-open paper/plastic wrappers. 221mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 600/case