Learn More
CELLTREAT Scientific Products 10l Inoculating Loop, Bulk Pack, Sterile
SDP
Catalog No. 50201984
Encompass_Preferred
CELLTREAT Scientific Products 10¿L Inoculating Loop. Flexible for easy access to small volume tubes and dishes. Color coded blue for easy identification. Gentle on delicate agar or other surfaces. 221mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 20/pack, 2000/case.
Catalog No. 50-201-984 Supplier CELLTREAT Scientific Products Supplier No. 229610
CELLTREAT Scientific Products 10¿L Inoculating Loop. Flexible for easy access to small volume tubes and dishes. Color coded blue for easy identification. Gentle on delicate agar or other surfaces. 221mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 20/pack, 2000/case.