Learn More
CELLTREAT Scientific Products 1l Inoculating Loop, Bulk Pack, Sterile
SDP
Catalog No. 50201982
Encompass_Preferred
CELLTREAT Scientific Products 1¿L Inoculating Loop. Flexible for easy access to small volume tubes and dishes. Color coded white for easy identification. Gentle on delicate agar or other surfaces. Available in peel-to-open paper/plastic wrappers. 218mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 20/pack, 2000/case.
Catalog No. 50-201-982 Supplier CELLTREAT Scientific Products Supplier No. 229601
CELLTREAT Scientific Products 1¿L Inoculating Loop. Flexible for easy access to small volume tubes and dishes. Color coded white for easy identification. Gentle on delicate agar or other surfaces. Available in peel-to-open paper/plastic wrappers. 218mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 20/pack, 2000/case.