Learn More
Electron Microscopy Sciences Inoculating Loop, Nichrome wire 12/pk
SDP
Catalog No. 5026715
Encompass_Preferred
Nichrome wire. An 8" aluminum handle with a 3" nichrome wire, 25 gauge. Loop O.D. is 4 mm; loop I.D. is 2.5 mm.
Catalog No. 50-267-15 Supplier Electron Microscopy Sciences Supplier No. 7295210
Nichrome wire. An 8" aluminum handle with a 3" nichrome wire, 25 gauge. Loop O.D. is 4 mm; loop I.D. is 2.5 mm.