Learn More
Electron Microscopy Sciences Inoculating Loops, 2x45 mm, 24 Gauge, 45 mm OAL
SDP
Catalog No. 5026724
Encompass_Preferred
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a "perfect circle".
Diameter: 2 mm
Wire: 24 gauge
OAL: 45 mm
Catalog No. 50-267-24 Supplier Electron Microscopy Sciences Supplier No. 6243302
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a "perfect circle".
Diameter: 2 mm
Wire: 24 gauge
OAL: 45 mm