Learn More
Electron Microscopy Sciences Inoculating Loops, 3x70 mm, 24 Gauge, 70 mm OAL
SDP
Catalog No. 5026721
Encompass_Preferred
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a "perfect circle".
Diameter: 3 mm
Wire: 24 gauge
OAL: 70 mm
Catalog No. 50-267-21 Supplier Electron Microscopy Sciences Supplier No. 6243603
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a "perfect circle".
Diameter: 3 mm
Wire: 24 gauge
OAL: 70 mm