Learn More
Chemglass Life Sciences 10µl Inoculating Loop, Blue
SDP
Catalog No. 501897589
Encompass
Loops and needles are made from high quality polypropylene and are flexible so¿stem can be bent to access small volume tubes and dishes. Color coded for easy¿identification. Vacuum-gas plasma surface treated. Packaged in peel-to-open paper¿plastic wrappers. Product is sterile (gamma irradiated).
Catalog No. 50-189-7589 Supplier Chemglass Life Sciences Supplier No. CGE2458010
Loops and needles are made from high quality polypropylene and are flexible so¿stem can be bent to access small volume tubes and dishes. Color coded for easy¿identification. Vacuum-gas plasma surface treated. Packaged in peel-to-open paper¿plastic wrappers. Product is sterile (gamma irradiated).