Learn More
Hach Company Loop, Inoculating, Wire
Catalog No. 501995427 Shop All Hach Company Products
Encompass
3" nichrome V wire, 25 gauge (0.018") with 3-mm diameter loop.
Catalog No. 50-199-5427 Supplier Hach Company Supplier No. 2112100
3" nichrome V wire, 25 gauge (0.018") with 3-mm diameter loop.