Learn More
Research Products International Corp Inoculating Loops 1ul, Blue, 1000/cs
SDP
Catalog No. 501977723
Encompass
High impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamper proof, zip-seal resealable bags.
Disposable inoculating loops and needles do not need flaming and thus eliminate the risk of infection due to aerosol formation of pathogenic substances. Cross-contamination due to improper sterilization is eliminated. They can be used under hoods without danger.
Catalog No. 50-197-7723 Supplier Research Products International Corp Supplier No. 246480
High impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamper proof, zip-seal resealable bags.
Disposable inoculating loops and needles do not need flaming and thus eliminate the risk of infection due to aerosol formation of pathogenic substances. Cross-contamination due to improper sterilization is eliminated. They can be used under hoods without danger.