Learn More
Andwin Scientific INOCULATING LOOPS DISPOSA
SDP
Catalog No. NC9652534
Encompass_Preferred
The 10 microliter Sterile Inoculating Loops feature flexibility to facilitate uniform and smooth streaking without damaging cell surface. This product is packed sterile in zippered resealable bags.
This item is not returnable. View return policy
Catalog No. NC9652534 Supplier Andwin Scientific Supplier No. 60872406
The 10 microliter Sterile Inoculating Loops feature flexibility to facilitate uniform and smooth streaking without damaging cell surface. This product is packed sterile in zippered resealable bags.