Learn More
Copan Diagnostics Inoculating Loops and Needles in Zip Lock Pack, Calibrated, Disposable
Calibrated 1 and 10μL plastic disposable inoculating loops and needles packaged in Zip-lock closure bags that reseal between uses to avoid contamination.
Supplier: Copan Diagnostics CSTS10
Description
- Re-sealable bag allows the user to open and close the loop pack to remove individual loops
- Disposable, sterile plastic loops and needles manufactured from soft-flexible plastic
- Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
- Ultra smooth surfaces allow easy streaking without cutting or gouging agar surface
- Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
- Color coded for easy product identification
- Packed in tamper-proof bags to guarantee sterility
- Certificate of calibration included in each case
Specifications
Yes | |
Plastic | |
Light Blue | |
Evans blue dye certificate of calibration | |
1000 | |
Yes |
10 μL | |
Sterile | |
Inoculating Loop, Flexible | |
201 mm | |
Zip lock resealable bags |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'