Learn More
Copan Diagnostics Inoculating Loops and Needles in Zip Lock Pack, Calibrated, Disposable
SDP
Catalog No. 23600970 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Volume:
1 μL
10 μL
Length (Metric) Handle:
199 mm
201 mm
Color:
Light Blue
Light Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Length (Metric) Handle Color
23-600-970 1 μL 199 mm Light Green
23-600-971 10 μL 201 mm Light Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-600-970 Supplier Copan Diagnostics Supplier No. CSTS1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Calibrated 1 and 10μL plastic disposable inoculating loops and needles packaged in Zip-lock closure bags that reseal between uses to avoid contamination.
  • Re-sealable bag allows the user to open and close the loop pack to remove individual loops
  • Disposable, sterile plastic loops and needles manufactured from soft-flexible plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultra smooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case
Specifications
Disposable Single-use
Volume 1 μL
Material Plastic
Sterility Sterile
Color Light Green
Product Type Inoculating Loop, Flexible
Includes Evans blue dye certificate of calibration
Length (Metric) Handle 199 mm
Quantity 1000
Packaging Type Zip lock resealable bags
Calibrated Yes