Learn More
GSC Go Science Crazy Inoculating Needle, Straight Nichrome Wire, Metal Handle
SCI_ED
Catalog No. S24602 Shop All GSC Products
Click to view available options
Handle Type:
Metal Handle
Catalog No. Handle Type
S24602 Metal Handle
1 options
Catalog No. S24602 Supplier GSC Go Science Crazy Supplier No. 42351310
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ideal for retrieving an inoculum from a culture or transferring microorganisms to streak plates or cultures. This straight needle is made out of nichrome wire. It is attached to a 6 in. long metal handle.
  • Nichrome wire inoculating needle with straight end
  • 6 inch metal handle
Specifications
Product Type Inoculating Needle, Straight Nichrome Wire
Handle Type Metal Handle
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov