Learn More
Electron Microscopy Sciences Inoculating Needle, 70 mm
SDP
Catalog No. 5026731
Encompass_Preferred
Made from 26 gauge BandS platinum wire with 15% iridium to provide rigidity with thinner wire for delicate work. Both ends are ground smooth.
Catalog No. 50-267-31 Supplier Electron Microscopy Sciences Supplier No. 6243201
Made from 26 gauge BandS platinum wire with 15% iridium to provide rigidity with thinner wire for delicate work. Both ends are ground smooth.