Learn More
Chemglass Life Sciences Inoculating Needle, Yellow
SDP
Catalog No. 501897590
Encompass
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification. Vacuum-gas plasma surface treated. Packaged in peel-to-open paper plastic wrappers. Product is sterile (gamma irradiated).
Catalog No. 50-189-7590 Supplier Chemglass Life Sciences Supplier No. CGE2458015
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification. Vacuum-gas plasma surface treated. Packaged in peel-to-open paper plastic wrappers. Product is sterile (gamma irradiated).