Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Inoculating Needle/Loops
GREENER_CHOICE

Catalog No. 03448343
Change view
Click to view available options
Volume:
1 μL
10 μL
Material:
ABS
Polypropylene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Material
03-448-343 1 μL Polypropylene
03-448-344 1 μL ABS
03-448-345 10 μL Polypropylene
03-448-346 10 μL ABS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-448-343 Supplier Heathrow Scientific™ Supplier No. HS81121A
Add to Cart
Add to Cart

Provide the choice to spread or pick colonies and cultures without the risk of contamination with Inoculating Needle/Loops.

  • Use loop end to uniformly streak a gel surface
  • Use needle end for removal of colonies
  • Lot-to-lot traceability
  • Sterile
  • Disposable

Specifications

Disposable Single-use
Material Polypropylene
Volume 1 μL
Length (Metric) 20 cm
Sterility Sterile
Length (English) 7.9 in.
Size Standard
Color Blue
Product Type Inoculating Loop and Needle, Flexible
Autoclavable Not Autoclavable
For Use With (Application) Spreading, Picking, and Transferring Colonies or Cultures
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.