Learn More
Chemglass Life Sciences Inoculating Needle, White, Individually Wrapped, Sterile, 400/Pack
SDP
Catalog No. 501897754
Encompass_Preferred
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification.PP material, hydrophilicUsed in microbiology experimentsDifferent colors for identificationFlexible handles for easier collection and inoculationSterilized by Ethylene OxideEasy-tear sterile individual packaging
Catalog No. 50-189-7754 Supplier Chemglass Life Sciences Supplier No. CGN2458015
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification.PP material, hydrophilicUsed in microbiology experimentsDifferent colors for identificationFlexible handles for easier collection and inoculationSterilized by Ethylene OxideEasy-tear sterile individual packaging