Learn More
Research Products International Corp Inoculating Loop 1ul, Blue Sterile, 20/pk, 500/cs
SDP
Catalog No. 502253018
Encompass
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polypropylene. Supplied in sealed packs. Packaged 20 per pack, 500 per case.
Smooth and flexible
Do not require flaming, eliminates risk of infection
This item is not returnable. View return policy
Catalog No. 50-225-3018 Supplier Research Products International Corp Supplier No. 161210A
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polypropylene. Supplied in sealed packs. Packaged 20 per pack, 500 per case.
Smooth and flexible
Do not require flaming, eliminates risk of infection