Learn More
Research Products International Corp Inoculating Needles Sterile ABS 20/pk 500/cs
SDP
Catalog No. 502253020
Encompass
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polystyrene for easy disposal after each use. Supplied in sealed packs with certification. Packaged 25 per pack, 500 per case.
This item is not returnable. View return policy
Catalog No. 50-225-3020 Supplier Research Products International Corp Supplier No. 161213A
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polystyrene for easy disposal after each use. Supplied in sealed packs with certification. Packaged 25 per pack, 500 per case.