Learn More
TARGETMOL CHEMICALS INC INOLITAZONE 5MG
SDP
Catalog No. 502581220
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Inolitazone (RS5444) a high-affinity PPAR(gamma) agonist. Inolitazone exhibits IC50 for growth inhibition is ~0.8 nM in vitro. purity: 99%
Catalog No. 50-258-1220 Supplier TARGETMOL CHEMICALS INC Supplier No. T155815MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Inolitazone (RS5444) a high-affinity PPAR(gamma) agonist. Inolitazone exhibits IC50 for growth inhibition is ~0.8 nM in vitro. purity: 99%