Learn More
Medchemexpress LLC Inolitazone dihydrochloride | 223132-38-5 | 98.5% | 575.51 | 100 MG
SDP
Catalog No. 5000361599
Encompass_Preferred
Inolitazone dihydrochloride is a novel high-affinity PPARγ agonist, demonstrating potent anti-proliferative activities both in vitro and in vivo, with an IC50 of 0.8 nM for growth inhibition. Its biological activity is dependent on PPARγ.
  • Upregulates the cell cycle kinase inhibitor p21WAF1/CIP1
  • Activates PPARγ:RXRα-dependent transcription at 10 nM
  • Specifically activates PPARγ, unlike PPARα or PPARδ
  • Exhibits a low EC50 of 1 nM for activating PPARγ in DRO cells
  • Potently inhibits growth with an IC50 of 0.8 nM
  • Shows additive antiproliferative activity when combined with paclitaxel
  • Inhibits tumor growth in a dose-responsive manner in vivo, including established DRO and ARO tumors
Catalog No. 50-003-61599 Supplier Medchemexpress LLC Supplier No. HY14792B100MG
Inolitazone dihydrochloride is a novel high-affinity PPARγ agonist, demonstrating potent anti-proliferative activities both in vitro and in vivo, with an IC50 of 0.8 nM for growth inhibition. Its biological activity is dependent on PPARγ.
  • Upregulates the cell cycle kinase inhibitor p21WAF1/CIP1
  • Activates PPARγ:RXRα-dependent transcription at 10 nM
  • Specifically activates PPARγ, unlike PPARα or PPARδ
  • Exhibits a low EC50 of 1 nM for activating PPARγ in DRO cells
  • Potently inhibits growth with an IC50 of 0.8 nM
  • Shows additive antiproliferative activity when combined with paclitaxel
  • Inhibits tumor growth in a dose-responsive manner in vivo, including established DRO and ARO tumors