Learn More
Aldevron Inorganic Pyrophosphatase, 1870ug
Catalog No. 503073872
Encompass_Preferred
1870ug dispense of Inorganic Pyrophosphatase. Inorganic Pyrophosphatase catalyzes hydrolysis of inorganic pyrophosphate, producing two molecules of phosphate.
Catalog No. 50-307-3872 Supplier Aldevron Supplier No. 91321870MCG
1870ug dispense of Inorganic Pyrophosphatase. Inorganic Pyrophosphatase catalyzes hydrolysis of inorganic pyrophosphate, producing two molecules of phosphate.