Learn More
Aldevron Inorganic Pyrophosphatase, 350ug
Catalog No. 503073880
Encompass_Preferred
350ug dispense of Inorganic Pyrophosphatase. Inorganic Pyrophosphatase catalyzes hydrolysis of inorganic pyrophosphate, producing two molecules of phosphate.
Catalog No. 50-307-3880 Supplier Aldevron Supplier No. 9132350MCG
350ug dispense of Inorganic Pyrophosphatase. Inorganic Pyrophosphatase catalyzes hydrolysis of inorganic pyrophosphate, producing two molecules of phosphate.