Learn More
Inorganic Ventures 147 CONDUCTIVITY 1L
SDP
Catalog No. NC2033128
Encompass_Preferred
Inorganic Ventures 147 UMHOS/CM CONDUCTIVITY STD Conductivity Standard (147 uS/cm at 25C). 1 Liter Bottle
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC2033128 Supplier Inorganic Ventures Supplier No. CON147251L
Inorganic Ventures 147 UMHOS/CM CONDUCTIVITY STD Conductivity Standard (147 uS/cm at 25C). 1 Liter Bottle
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search