Learn More
Inorganic Ventures Inorganic Ventures Working Buffer Reagent, Nitrate/Nitrite Analysis, Cadmium Coil Reduction, 125 mL
SDP
Catalog No. NC3912971
Encompass
This working buffer reagent is specifically formulated for nitrate and nitrite analysis using the cadmium coil reduction method. It is designed to ensure optimal performance in analytical procedures requiring the reduction of nitrate to nitrite.
  • Optimized for nitrate and nitrite analysis via cadmium coil reduction
  • Compatible with seal epa-114, epa-126, epa-127, epa 353.2, and standard methods 4500-no3 f
  • Volume of 125 ml
  • Guaranteed stability of 12 months
Catalog No. NC3912971 Supplier Inorganic Ventures Supplier No. SEALANALYTICALAQNOXCADBUF
This working buffer reagent is specifically formulated for nitrate and nitrite analysis using the cadmium coil reduction method. It is designed to ensure optimal performance in analytical procedures requiring the reduction of nitrate to nitrite.
  • Optimized for nitrate and nitrite analysis via cadmium coil reduction
  • Compatible with seal epa-114, epa-126, epa-127, epa 353.2, and standard methods 4500-no3 f
  • Volume of 125 ml
  • Guaranteed stability of 12 months