Learn More
ERA WATERS CORP INORGANIS DISINFECTION #2,QC
SDP
Catalog No. NC0344041
Encompass
Inorganic Disinfection 2 QC One 23 mL screw-cap vial yields up to 4 liters after dilution.
This item is not returnable. View return policy
Catalog No. NC0344041 Supplier ERA WATERS CORP Supplier No. 5262
Inorganic Disinfection 2 QC One 23 mL screw-cap vial yields up to 4 liters after dilution.