Learn More
Enzo Life Sciences iNOS polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKASNO001D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 1131-1144 of mouse iNOS. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat, Bovine. Applications: IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7317