Learn More
ECM Bioscience iNOS Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience NK6450
Encompass_Preferred
The inducible nitric oxide synthase (iNOS) phospho-regulation antibody sampler kit can be used to examine Tyr-1055 phosphorylation in iNOS. The kit also includes a mouse monoclonal and rabbit polyclonal antibody to monitor total expression levels of iNOS, as well as HRP-conjugated secondary reagents. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8759