Learn More
Cayman Chemical InosIn 5-trIphosphatesodI 50mg
SDP
Catalog No. 502964476
Encompass_Preferred
A nucleoside triphosphate; alternative substrate for ATPases and GTPases; has been used to study activation and binding kinetics of nucleoside interaction with various ATPases and GTPases
Catalog No. 50-296-4476 Supplier Cayman Chemical Supplier No. 2306350 MG
A nucleoside triphosphate; alternative substrate for ATPases and GTPases; has been used to study activation and binding kinetics of nucleoside interaction with various ATPases and GTPases