Learn More
Medchemexpress LLC Inosine-13C5 | 58-63-9 | 99.9% | 273.19 | 5 MG
SDP
Catalog No. 5000427692
Encompass_Preferred
Inosine-13C5 is the 13C5 labeled version of Inosine. Inosine is a naturally occurring purine nucleoside produced from adenosine catabolism. It exhibits anti-inflammatory, antinociceptive, immunomodulatory, and neuroprotective effects, acting as an agonist for adenosine A1 (A1R) and A2A (A2AR) receptors.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like carbon-13 are incorporated into drug molecules as tracers for quantitation in drug development.
  • Dose-dependently stimulates cAMP production through A2AR.
  • Dose-dependently induces hA2AR-mediated ERK1/2 phosphorylation.
  • Reduces oxidative stress in MES 23.5 cells cultured with astrocytes at 100 μM for 24 hours.
Catalog No. 50-004-27692 Supplier Medchemexpress LLC Supplier No. HYN0092S25MG
Inosine-13C5 is the 13C5 labeled version of Inosine. Inosine is a naturally occurring purine nucleoside produced from adenosine catabolism. It exhibits anti-inflammatory, antinociceptive, immunomodulatory, and neuroprotective effects, acting as an agonist for adenosine A1 (A1R) and A2A (A2AR) receptors.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like carbon-13 are incorporated into drug molecules as tracers for quantitation in drug development.
  • Dose-dependently stimulates cAMP production through A2AR.
  • Dose-dependently induces hA2AR-mediated ERK1/2 phosphorylation.
  • Reduces oxidative stress in MES 23.5 cells cultured with astrocytes at 100 μM for 24 hours.