Learn More
Medchemexpress LLC Inosine-13C5 | 58-63-9 | MFCD00066770 | 99.9% | 273.19 | C10H12N4O5 (13C5 labeled) | 10 MG
SDP
Catalog No. 5000295974
Encompass_Preferred
Inosine-13C5 is a 13C5 isotopically labeled inosine supplied as a powder for use as an analytical or internal standard in metabolomics and biochemical assays. It is high-purity and intended for research use with recommended solvent and storage conditions to preserve stability.
  • Isotopically labeled (13C5) internal standard for metabolomics.
  • High purity: 99.9%.
  • Available in small pack sizes suitable for analytical workflows.
  • Powder form; soluble in DMSO at 10 mg/mL.
  • Stable when stored as powder at -20°C; solution stability at -80°C for up to 6 months.
Catalog No. 50-002-95974 Supplier Medchemexpress LLC Supplier No. HYN0092S210MG
Inosine-13C5 is a 13C5 isotopically labeled inosine supplied as a powder for use as an analytical or internal standard in metabolomics and biochemical assays. It is high-purity and intended for research use with recommended solvent and storage conditions to preserve stability.
  • Isotopically labeled (13C5) internal standard for metabolomics.
  • High purity: 99.9%.
  • Available in small pack sizes suitable for analytical workflows.
  • Powder form; soluble in DMSO at 10 mg/mL.
  • Stable when stored as powder at -20°C; solution stability at -80°C for up to 6 months.