Learn More
Medchemexpress LLC Inosine-15N4 | 204634-01-5 | C10H1215N4O5 | 1 MG
SDP
Catalog No. 5000375714
Encompass_Preferred
Inosine-15N4 is the 15N labeled Inosine, primarily used as a tracer and internal standard for quantitative analysis in drug development. This compound is valuable for techniques such as NMR, GC-MS, or LC-MS by incorporating stable heavy isotopes into drug molecules.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes into drug molecules
Catalog No. 50-003-75714 Supplier Medchemexpress LLC Supplier No. HY150792S1MG
Inosine-15N4 is the 15N labeled Inosine, primarily used as a tracer and internal standard for quantitative analysis in drug development. This compound is valuable for techniques such as NMR, GC-MS, or LC-MS by incorporating stable heavy isotopes into drug molecules.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes into drug molecules