Learn More
Selleck Chemical LLC Inosine 25mg 58-63-9 NSC 20262, INO 495
SDP
Supplier:  Selleck Chemical LLC S244225MG
Encompass_Preferred
Inosine (NSC 20262, INO 495) is a nucleoside that is formed when hypoxanthine is attached to a ribose ring via a β-N9-glycosidic bond. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8255