Learn More
Medchemexpress LLC Inosine 5′-diphosphate sodium | 81012-88-6 | 98.17% | 512.15 | 250 MG
SDP
Catalog No. 5000366102
Encompass_Preferred
Inosine 5′-diphosphate sodium is a purine ribonucleoside 5′-diphosphate with inosine as the nucleobase. It participates in intracellular energy metabolism and signal transduction processes.
  • Purine ribonucleoside 5′-diphosphate with inosine as the nucleobase
  • Participates in intracellular energy metabolism
  • Participates in signal transduction processes
  • Functions as an endogenous metabolite
  • Classified as a nucleotide analog
  • Used in research for respiratory disease
  • Used in research for metabolic or endocrine disease
  • Used in research for pulmonary disease
  • Used in research for lung fibrosis
Catalog No. 50-003-66102 Supplier Medchemexpress LLC Supplier No. 250MGHYW749117
Inosine 5′-diphosphate sodium is a purine ribonucleoside 5′-diphosphate with inosine as the nucleobase. It participates in intracellular energy metabolism and signal transduction processes.
  • Purine ribonucleoside 5′-diphosphate with inosine as the nucleobase
  • Participates in intracellular energy metabolism
  • Participates in signal transduction processes
  • Functions as an endogenous metabolite
  • Classified as a nucleotide analog
  • Used in research for respiratory disease
  • Used in research for metabolic or endocrine disease
  • Used in research for pulmonary disease
  • Used in research for lung fibrosis