Learn More
Medchemexpress LLC Inosine 5'-diphosphate sodium | 81012-88-6 | 98.17% | 512.15 | 500 MG
SDP
Catalog No. 5000365882
Encompass_Preferred
Inosine 5'-diphosphate sodium is a purine ribonucleoside 5'-diphosphate with inosine as the nucleobase. It participates in intracellular energy metabolism and signal transduction processes.
  • Purine ribonucleoside 5'-diphosphate
  • Contains inosine as the nucleobase
  • Participates in intracellular energy metabolism
  • Involved in signal transduction processes
Catalog No. 50-003-65882 Supplier Medchemexpress LLC Supplier No. 500MGHYW749117
Inosine 5'-diphosphate sodium is a purine ribonucleoside 5'-diphosphate with inosine as the nucleobase. It participates in intracellular energy metabolism and signal transduction processes.
  • Purine ribonucleoside 5'-diphosphate
  • Contains inosine as the nucleobase
  • Participates in intracellular energy metabolism
  • Involved in signal transduction processes