Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inosine 5'-Monophosphate Barium Salt Hydrate 95.0+%, TCI America™
SDP

Catalog No. I0039100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0039100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0039100MG Supplier TCI America Supplier No. I0039100MG

Chemical Identifiers

CAS 3249-92-1
Molecular Formula C10H13BaN4O8P
Molecular Weight (g/mol) 485.535
MDL Number MFCD00067321
InChI Key JKIAOYFSNWPEEW-MCDZGGTQSA-N
Synonym Barium Inosinate, Inosic Acid Barium Salt, 5′C-Inosinic Acid Barium Salt, 5′C-IMP Ba
PubChem CID 131877769
IUPAC Name barium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate
SMILES C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)COP(=O)(O)O)O)O.[Ba]

Specifications

Color White
Quantity 100 mg
UN Number 1564
Formula Weight 483.52
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Inosine 5′-Monophosphate Barium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.