Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Inosine-5'-monophosphate disodium salt hydrate

Catalog No. AAJ6195914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6195914 25 g
AAJ6195906 5 g
AAJ6195922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6195914 Supplier Thermo Scientific Chemicals Supplier No. J6195914
Add to Cart

CAS: 352195-40-5 | C10H11N4Na2O8P | 392.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352195-40-5
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00036201,MFCD00036201
InChI Key AANLCWYVVNBGEE-IDIVVRGQSA-L
Synonym disodium 5'-inosinate
PubChem CID 123134061
IUPAC Name [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O

Specifications

Color White
Quantity 25 g
Formula Weight 392.17 (Anhydrous)
Physical Form Crystalline Powder
Chemical Name or Material Inosine-5'-monophosphate disodium salt hydrate
EINECSNumber 225-146-4
RTECSNumber NM7519500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.