Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inosine 5'-Monophosphate Disodium Salt, Hydrate, Spectrum™ Chemical
SDP

Catalog No. 18620945 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-945 5 g
18-620-946 25 g
18-620-947 100 g
18-620-948 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-945 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I30415GM
Add to Cart
Add to Cart

Synonym: 5'-Inosinic Acid Disodium Salt Hydrate, Disodium 5'-Inosinate Monohydrate, 5'-Inosinic Acid Disodium Hydrate

Chemical Identifiers

CAS 4691-65-0

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.