Learn More
Medchemexpress LLC Inosine-5'-monophosphate disodium salt octahydrate | 20813-76-7 | >98.0% | 536.29 | 25 G
SDP
Catalog No. 5000366795
Encompass_Preferred
Inosine-5'-monophosphate disodium salt octahydrate is a purine nucleotide that can be used as an umami tastant as well as a precursor of GMP and AMP.
  • Endogenous metabolite
  • Purine nucleotide
  • Can be used as an umami tastant
  • Precursor of GMP and AMP
Catalog No. 50-003-66795 Supplier Medchemexpress LLC Supplier No. 25GHYW010759
Inosine-5'-monophosphate disodium salt octahydrate is a purine nucleotide that can be used as an umami tastant as well as a precursor of GMP and AMP.
  • Endogenous metabolite
  • Purine nucleotide
  • Can be used as an umami tastant
  • Precursor of GMP and AMP