Learn More
Apexbio Technology LLC INOSINE-5-PO4 DINA-200mg
SDP
Catalog No. 5000897507
Encompass_Preferred
Inosine 5 -monophosphate disodium salt (CAS No 352195-40-5) is a nucleotide derivative often utilized in biomedical research for its role in purine metabolism It originates from the metabolic processes of nucleic acids and participates in vital biochemical pathways including the biosynthesis of RNA and DNA This compound has demonstrated efficacy in various in vitro activities typically within the nanomolar to low micromolar range Its primary applications include studies involving cell models and animal experiments as well as drug screening programs Experimental concentrations typically depend on the specific research design allowing for adaptability in various experimental contexts It is widely used by researchers to explore its effects on cellular functions and to investigate potential therapeutic avenues in models of disease
Catalog No. 50-008-97507 Supplier Apexbio Technology LLC Supplier No. C6982200
Inosine 5 -monophosphate disodium salt (CAS No 352195-40-5) is a nucleotide derivative often utilized in biomedical research for its role in purine metabolism It originates from the metabolic processes of nucleic acids and participates in vital biochemical pathways including the biosynthesis of RNA and DNA This compound has demonstrated efficacy in various in vitro activities typically within the nanomolar to low micromolar range Its primary applications include studies involving cell models and animal experiments as well as drug screening programs Experimental concentrations typically depend on the specific research design allowing for adaptability in various experimental contexts It is widely used by researchers to explore its effects on cellular functions and to investigate potential therapeutic avenues in models of disease