Learn More
Selleck Chemical LLC Inosine 5-triphosphate trisodium salt 25mg 35908-31-7
SDP
Supplier:  Selleck Chemical LLC S610325MG
Encompass_Preferred
Inosine 5?-triphosphate (ITP) is used in studies on the impact of deamination of ATP and GTP by various enzymes and chemical processes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1896849