Learn More
Medchemexpress LLC Inosine pranobex | 36703-88-5 | 98.1% | 1115.23 g/mol | C52H78N10O17 | 100 MG
SDP
Catalog No. 5000244484
Encompass_Preferred
Inosine pranobex is an orally active immunomodulator supplied for research and analytical use. It exhibits broad-spectrum antiviral activity and is commonly used in virology and immunology studies to evaluate antiviral and immune-modulating effects in vitro.
  • Orally active immunomodulator.
  • Broad-spectrum antiviral activity against multiple DNA and RNA viruses.
  • Research-grade purity: 98.1%.
  • Molecular weight 1115.23 g/mol.
  • Available as solids (for example, 100 mg and larger) and as 10 mM solutions in DMSO.
  • Intended for in vitro and analytical research applications.
Catalog No. 50-002-44484 Supplier Medchemexpress LLC Supplier No. HY107801100MG
Inosine pranobex is an orally active immunomodulator supplied for research and analytical use. It exhibits broad-spectrum antiviral activity and is commonly used in virology and immunology studies to evaluate antiviral and immune-modulating effects in vitro.
  • Orally active immunomodulator.
  • Broad-spectrum antiviral activity against multiple DNA and RNA viruses.
  • Research-grade purity: 98.1%.
  • Molecular weight 1115.23 g/mol.
  • Available as solids (for example, 100 mg and larger) and as 10 mM solutions in DMSO.
  • Intended for in vitro and analytical research applications.