Learn More
Medchemexpress LLC Inosine pranobex | 36703-88-5 | MFCD05662374 | >98.0% | 1115.2 g/mol | C52H78N10O17 | 5 G
SDP
Catalog No. 5000242424
Encompass_Preferred
Inosine pranobex is an oral immunomodulator and antiviral salt complex supplied as a white to off-white powder for research and analytical use. The compound has CAS 36703-88-5, molecular formula C52H78N10O17, and a molar mass of approximately 1115.2 g/mol, and is used in studies of antiviral activity and immune modulation.
  • Oral immunomodulator with broad-spectrum antiviral activity.
  • Supplied as a powder suitable for formulation or analytical applications.
  • Molecular formula C52H78N10O17 and molecular weight 1115.2 g/mol.
  • Typical supplier-reported purity is >98.0% by HPLC.
  • Available in multiple pack sizes, including a 5 G research pack.
Catalog No. 50-002-42424 Supplier Medchemexpress LLC Supplier No. HY1078015G
Inosine pranobex is an oral immunomodulator and antiviral salt complex supplied as a white to off-white powder for research and analytical use. The compound has CAS 36703-88-5, molecular formula C52H78N10O17, and a molar mass of approximately 1115.2 g/mol, and is used in studies of antiviral activity and immune modulation.
  • Oral immunomodulator with broad-spectrum antiviral activity.
  • Supplied as a powder suitable for formulation or analytical applications.
  • Molecular formula C52H78N10O17 and molecular weight 1115.2 g/mol.
  • Typical supplier-reported purity is >98.0% by HPLC.
  • Available in multiple pack sizes, including a 5 G research pack.