Learn More
Abcam Inosine, Purine nucleoside, 100MG
Catalog No. 502707334
Encompass_Preferred
MW 268.23 g/mol, Purity >99%. Purine nucleoside and adenosine metabolite. Cell-permeable. Displays neuroprotective and immunomodulatory effects. Potent vasodilator and axon growth stimulator.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7334 Supplier Abcam Supplier No. AB120724100MG
MW 268.23 g/mol, Purity >99%. Purine nucleoside and adenosine metabolite. Cell-permeable. Displays neuroprotective and immunomodulatory effects. Potent vasodilator and axon growth stimulator.
The product is subject to the following: Abcam Restricted Use Statement